COMH4211AD97 Combi-Blocks, Inc.

5-Bromo-2-(2,2,2-trifluoroethoxy)aniline

  • MDL number MFCD11132420
  • Cas Number : 1019121-52-8
  • Molecular Weight : 270.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2191

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2191
Availability details for
Quantity
  • Availability
  • Add to Cart