COMH4211AD9C Combi-Blocks, Inc.

3-(4-Hydroxy-pyrazol-1-ylmethyl)-piperidine-1-carboxylic acid tert-butyl ester

  • MDL number MFCD31561595
  • Cas Number : 2270906-53-9
  • Molecular Weight : 281.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2207

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2207
Availability details for
Quantity
  • Availability
  • Add to Cart