COMH4211AD9D Combi-Blocks, Inc.

Calcium (3s,5r)-7-(2-(4-fluorophenyl)-5-isopropyl-3-phenyl-4-(phenylcarbamoyl)-1h-pyrrol-1-yl)-3,5-dihydroxyheptanoate

  • MDL number MFCD24682768
  • Cas Number : 887196-25-0
  • Molecular Weight : 1155.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2208

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2208
Availability details for
Quantity
  • Availability
  • Add to Cart