COMH4211AD9F Combi-Blocks, Inc.

1-(3-Cyclopropylmethoxy-benzyl)-piperazine

  • MDL number MFCD31561468
  • Cas Number : 1891508-55-6
  • Molecular Weight : 246.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2217

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2217
Availability details for
Quantity
  • Availability
  • Add to Cart