COMH4211ADA0 Combi-Blocks, Inc.

(4-Hydroxy-3,5-dimethyl-pyrazol-1-yl)-acetic acid methyl ester

  • MDL number MFCD31583064
  • Cas Number : 2270907-35-0
  • Molecular Weight : 184.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2218

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2218
Availability details for
Quantity
  • Availability
  • Add to Cart