COMH4211ADA2 Combi-Blocks, Inc.

3-Methoxypropionic acid hydrazide

  • MDL number MFCD03789158
  • Cas Number : 21920-89-8
  • Molecular Weight : 118.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2221

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2221
Availability details for
Quantity
  • Availability
  • Add to Cart