COMH4211ADA3 Combi-Blocks, Inc.

1H-Pyrazole, 1-([1,1'-biphenyl]-3-ylmethyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-

  • MDL number MFCD28718221
  • Cas Number : 1430751-24-8
  • Molecular Weight : 360.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2224

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2224
Availability details for
Quantity
  • Availability
  • Add to Cart