COMH4211ADA4 Combi-Blocks, Inc.

L-Norvalinamide, n-[(phenylmethoxy)carbonyl]-l-alanyl-

  • MDL number MFCD00025513
  • Cas Number : 197237-15-3
  • Molecular Weight : 321.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2228

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2228
Availability details for
Quantity
  • Availability
  • Add to Cart