COMH4211ADA5 Combi-Blocks, Inc.

(Cyclopropylmethyl)[(2,6-dichlorophenyl)methyl]amine

  • MDL number MFCD11140441
  • Cas Number : 1019553-89-9
  • Molecular Weight : 230.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2230

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2230
Availability details for
Quantity
  • Availability
  • Add to Cart