COMH4211ADA6 Combi-Blocks, Inc.

4-Acetyl-2-methyl-benzamide

  • MDL number MFCD24643465
  • Cas Number : 1095275-06-1
  • Molecular Weight : 177.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2231

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2231
Availability details for
Quantity
  • Availability
  • Add to Cart