COMH4211ADA7 Combi-Blocks, Inc.

3-[2-(2-Benzyloxycarbonylamino-ethoxy)-ethoxy]-propionic acid 2,5-dioxo-pyrrolidin-1-yl ester

  • MDL number MFCD31381628
  • Cas Number : 2203140-35-4
  • Molecular Weight : 408.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2232

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2232
Availability details for
Quantity
  • Availability
  • Add to Cart