COMH4211ADAC Combi-Blocks, Inc.

4-Amino-n-(4-bromophenyl)piperidine-1-carboxamide

  • MDL number MFCD30486591
  • Cas Number : 2044713-15-5
  • Molecular Weight : 298.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2243

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2243
Availability details for
Quantity
  • Availability
  • Add to Cart