COMH4211ADAE Combi-Blocks, Inc.

2-[(4-Methoxyphenyl)methoxy]aniline hydrochloride

  • MDL number MFCD21090498
  • Cas Number : 1353502-05-2
  • Molecular Weight : 265.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2253

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2253
Availability details for
Quantity
  • Availability
  • Add to Cart