COMH4211ADAF Combi-Blocks, Inc.

3-(4-Fluorophenyl)pyridin-2-amine

  • MDL number MFCD14698771
  • Cas Number : 1214333-67-1
  • Molecular Weight : 188.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2257

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2257
Availability details for
Quantity
  • Availability
  • Add to Cart