COMH4211ADB0 Combi-Blocks, Inc.

(4-Chloro-2-fluoro-benzyl)-isobutyl-amine

  • MDL number MFCD12815878
  • Cas Number : 1179950-73-2
  • Molecular Weight : 215.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2258

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2258
Availability details for
Quantity
  • Availability
  • Add to Cart