COMH4211ADB1 Combi-Blocks, Inc.

2-[(4-Methylpiperidin-1-yl)methyl]aniline

  • MDL number MFCD09815298
  • Cas Number : 1016752-31-0
  • Molecular Weight : 204.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2259

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2259
Availability details for
Quantity
  • Availability
  • Add to Cart