COMH4211ADB2 Combi-Blocks, Inc.

2-Methoxy-4'-(trifluoromethyl)biphenyl-4-carbonitrile

  • MDL number MFCD29110831
  • Cas Number : 1383803-31-3
  • Molecular Weight : 277.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2261

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2261
Availability details for
Quantity
  • Availability
  • Add to Cart