COMH4211ADB3 Combi-Blocks, Inc.

4-(Tetrahydro-pyran-4-ylmethyl)-benzoic acid

  • MDL number MFCD31614028
  • Cas Number : 1781028-66-7
  • Molecular Weight : 220.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2264

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2264
Availability details for
Quantity
  • Availability
  • Add to Cart