COMH4211ADB7 Combi-Blocks, Inc.

4-Acetyl-2-methylbenzoic acid

  • MDL number MFCD20639525
  • Cas Number : 55860-35-0
  • Molecular Weight : 178.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2276

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2276
Availability details for
Quantity
  • Availability
  • Add to Cart