COMH4211ADBB Combi-Blocks, Inc.

3-Chloro-4-cyclopropylmethoxy-benzylamine

  • MDL number MFCD16045358
  • Cas Number : 1248810-05-0
  • Molecular Weight : 211.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2281

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2281
Availability details for
Quantity
  • Availability
  • Add to Cart