COMH4211ADBC Combi-Blocks, Inc.

Ethyl 3-(6-methylpyridin-2-yl)-3-oxopropanoate

  • MDL number MFCD11974620
  • Cas Number : 150401-96-0
  • Molecular Weight : 207.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2282

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2282
Availability details for
Quantity
  • Availability
  • Add to Cart