COMH4211ADBD Combi-Blocks, Inc.

2-Bromo-6-(pyrrolidin-1-ylmethyl)pyridine

  • MDL number MFCD22199904
  • Cas Number : 1113040-40-6
  • Molecular Weight : 241.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2284

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2284
Availability details for
Quantity
  • Availability
  • Add to Cart