COMH4211ADC0 Combi-Blocks, Inc.

1-(2-Bromo-4-trifluoromethyl-phenyl)-ethylamine, hydrochloride

  • MDL number MFCD31539522
  • Cas Number : 2204562-89-8
  • Molecular Weight : 304.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2290

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2290
Availability details for
Quantity
  • Availability
  • Add to Cart