COMH4211ADC3 Combi-Blocks, Inc.

1-(3-Chloro-benzyl)-1h-pyrazol-4-ol

  • MDL number MFCD31561490
  • Cas Number : 1593233-73-8
  • Molecular Weight : 208.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2298

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2298
Availability details for
Quantity
  • Availability
  • Add to Cart