COMH4211ADC4 Combi-Blocks, Inc.

4-Amino-piperidine-1-carboxylic acid benzylamide hydrochloride

  • MDL number MFCD31557848
  • Cas Number : 2203714-65-0
  • Molecular Weight : 269.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2299

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2299
Availability details for
Quantity
  • Availability
  • Add to Cart