COMH4211ADCA Combi-Blocks, Inc.

4-(2-Chloropyrimidin-4-ylthio)benzenamine

  • MDL number MFCD31614112
  • Cas Number : 1042129-34-9
  • Molecular Weight : 237.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2308

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2308
Availability details for
Quantity
  • Availability
  • Add to Cart