COMH4211ADCC Combi-Blocks, Inc.

4-Amino-2-(cyclopropylmethoxy)benzamide

  • MDL number MFCD19640756
  • Cas Number : 1343847-18-6
  • Molecular Weight : 206.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2311

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2311
Availability details for
Quantity
  • Availability
  • Add to Cart