COMH4211ADCD Combi-Blocks, Inc.

Toluene-4-sulfonic acid 9-iodo-nonyl ester

  • MDL number MFCD31381689
  • Cas Number : 2206610-54-8
  • Molecular Weight : 424.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2313

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2313
Availability details for
Quantity
  • Availability
  • Add to Cart