COMH4211ADCE Combi-Blocks, Inc.

Ethyl (cyclopropylcarbamoyl)formate

  • MDL number MFCD10018323
  • Cas Number : 722486-66-0
  • Molecular Weight : 157.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2316

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2316
Availability details for
Quantity
  • Availability
  • Add to Cart