COMH4211ADCF Combi-Blocks, Inc.

1-(2-Aminoethoxy)-4-(trifluoromethyl)benzene

  • MDL number MFCD06248868
  • Cas Number : 681482-61-1
  • Molecular Weight : 205.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2317

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2317
Availability details for
Quantity
  • Availability
  • Add to Cart