COMH4211ADD0 Combi-Blocks, Inc.

4-Bromo-n-cyclopropyl-3-fluorobenzamide

  • MDL number MFCD16084871
  • Cas Number : 1249139-26-1
  • Molecular Weight : 258.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2319

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2319
Availability details for
Quantity
  • Availability
  • Add to Cart