COMH4211ADD2 Combi-Blocks, Inc.

2-Bromo-5-ethoxy-4-methylbenzaldehyde

  • MDL number MFCD26402585
  • Cas Number : 1350759-98-6
  • Molecular Weight : 243.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-bromo-5-ethoxy-4-methylbenzaldehyde
Inchi Code 1S/C10H11BrO2/c1-3-13-10-5-8(6-12)9(11)4-7(10)2/h4-6H,3H2,1-2H3
Inchi Key FUKUBLDYCHWDOM-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2321

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2321
Availability details for
Quantity
  • Availability
  • Add to Cart