COMH4211ADD3 Combi-Blocks, Inc.

3-(4,4-Difluoro-piperidin-1-yl)-propylamine

  • MDL number MFCD13184097
  • Cas Number : 869493-53-8
  • Molecular Weight : 178.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2323

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2323
Availability details for
Quantity
  • Availability
  • Add to Cart