COMH4211ADD4 Combi-Blocks, Inc.

1-Bromo-3-(but-3-en-1-yloxy)benzene

  • MDL number MFCD20391451
  • Cas Number : 1065169-60-9
  • Molecular Weight : 227.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-bromo-3-(3-butenyloxy)benzene
Inchi Code 1S/C10H11BrO/c1-2-3-7-12-10-6-4-5-9(11)8-10/h2,4-6,8H,1,3,7H2
Inchi Key IOFXHPULKXRKOP-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2324

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2324
Availability details for
Quantity
  • Availability
  • Add to Cart