COMH4211ADD5 Combi-Blocks, Inc.

Diphenyl ((trifluoromethyl)sulfonyl)phosphoramidate

  • MDL number MFCD29917433
  • Cas Number : 1009647-84-0
  • Molecular Weight : 381.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name diphenyl ((trifluoromethyl)sulfonyl)phosphoramidate
Inchi Code 1S/C13H11F3NO5PS/c14-13(15,16)24(19,20)17-23(18,21-11-7-3-1-4-8-11)22-12-9-5-2-6-10-12/h1-10H,(H,17,18)
Inchi Key GTRQXZJVYZSIKU-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2326

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2326
Availability details for
Quantity
  • Availability
  • Add to Cart