COMH4211ADD6 Combi-Blocks, Inc.

5-Bromo-4-methyl-pyridine-2-carboxylic acid ethylamide

  • MDL number MFCD31613905
  • Cas Number : 1403330-31-3
  • Molecular Weight : 243.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2335

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2335
Availability details for
Quantity
  • Availability
  • Add to Cart