COMH4211ADD8 Combi-Blocks, Inc.

2-[2-(4,4,5,5-Tetramethyl-[1,3,2]dioxaborolan-2-yl)-phenoxy]-ethyl-carbamic acid tert-butyl ester

  • MDL number MFCD31614106
  • Cas Number : 1808251-82-2
  • Molecular Weight : 363.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2339

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2339
Availability details for
Quantity
  • Availability
  • Add to Cart