COMH4211ADD9 Combi-Blocks, Inc.

6-(2-Fluoro-4-formyl-phenoxy)-nicotinonitrile

  • MDL number MFCD31618954
  • Cas Number : 676496-07-4
  • Molecular Weight : 242.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2341

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2341
Availability details for
Quantity
  • Availability
  • Add to Cart