COMH4211ADDA Combi-Blocks, Inc.

4-(3-Hydroxypropoxy)-3-methylbenzoic acid

  • MDL number MFCD16852898
  • Cas Number : 1273745-67-7
  • Molecular Weight : 210.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-(3-hydroxypropoxy)-3-methylbenzoic acid
Inchi Code 1S/C11H14O4/c1-8-7-9(11(13)14)3-4-10(8)15-6-2-5-12/h3-4,7,12H,2,5-6H2,1H3,(H,13,14)
Inchi Key AIQBHUNKAPLXQB-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2344

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2344
Availability details for
Quantity
  • Availability
  • Add to Cart