COMH4211ADDC Combi-Blocks, Inc.

1-(2-Chloro-4-iodo-phenyl)-ethylamine hydrochloride

  • MDL number MFCD31579432
  • Molecular Weight : 318.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2354

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2354
Availability details for
Quantity
  • Availability
  • Add to Cart