COMH4211ADDD Combi-Blocks, Inc.

1-(3,4-Dimethylbenzoyl)piperidine-4-carboxylic acid

  • MDL number MFCD09810669
  • Cas Number : 1017381-41-7
  • Molecular Weight : 261.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2356

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2356
Availability details for
Quantity
  • Availability
  • Add to Cart