COMH4211ADDE Combi-Blocks, Inc.

5-Chloro-2-phenylmethanesulfonylaniline

  • MDL number MFCD23067572
  • Cas Number : 74462-42-3
  • Molecular Weight : 281.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2360

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2360
Availability details for
Quantity
  • Availability
  • Add to Cart