COMH4211ADDF Combi-Blocks, Inc.

Pentanamide, 2-amino-n-[(4-fluorophenyl)methyl]-3-methyl-, (2s,3s)-

  • MDL number MFCD13245000
  • Cas Number : 1056678-67-1
  • Molecular Weight : 238.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2362

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2362
Availability details for
Quantity
  • Availability
  • Add to Cart