COMH4211ADE0 Combi-Blocks, Inc.

(2R)-3-Methyl-2-([(prop-2-en-1-yloxy)carbonyl]amino)butanoic

  • MDL number MFCD21252361
  • Cas Number : 132402-46-1
  • Molecular Weight : 201.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2368

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2368
Availability details for
Quantity
  • Availability
  • Add to Cart