COMH4211ADE1 Combi-Blocks, Inc.

3-Allyloxyphenylacetic acid

  • MDL number MFCD14696454
  • Cas Number : 156945-58-3
  • Molecular Weight : 192.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2375

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2375
Availability details for
Quantity
  • Availability
  • Add to Cart