COMH4211ADE2 Combi-Blocks, Inc.

2-(tert-Butylsulfanyl)-6-chloropyridine

  • MDL number MFCD18368387
  • Cas Number : 122733-89-5
  • Molecular Weight : 201.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2376

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2376
Availability details for
Quantity
  • Availability
  • Add to Cart