COMH4211ADE3 Combi-Blocks, Inc.

4-Chloro-3-(2,2,2-trifluoroethoxy)aniline

  • MDL number MFCD19625021
  • Cas Number : 1342115-81-4
  • Molecular Weight : 225.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2377

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2377
Availability details for
Quantity
  • Availability
  • Add to Cart