COMH4211ADE5 Combi-Blocks, Inc.

Benzenemethanamine, n-(2-propen-1-yloxy)-

  • MDL number MFCD31561559
  • Cas Number : 54615-13-3
  • Molecular Weight : 163.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2379

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2379
Availability details for
Quantity
  • Availability
  • Add to Cart