COMH4211ADE6 Combi-Blocks, Inc.

5-Bromo-2-isopropoxy-4-methyl-pyrimidine

  • MDL number MFCD31583179
  • Cas Number : 1896867-78-9
  • Molecular Weight : 231.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2380

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2380
Availability details for
Quantity
  • Availability
  • Add to Cart