COMH4211ADEA Combi-Blocks, Inc.

5-Chloro-2-[(2-methylpropyl)sulfanyl]aniline

  • MDL number MFCD11128891
  • Cas Number : 1019388-42-1
  • Molecular Weight : 215.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2390

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2390
Availability details for
Quantity
  • Availability
  • Add to Cart